RIE156329910 Rieke Metals, Inc.

6-oxo-6-(2-trifluoromethylphenyl)hexanoic acid

  • MDL number MFCD02260913
  • Molecular Weight : 274.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7139D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7139D
Availability details for
Quantity
  • Availability
  • Add to Cart