RIE156329912 Rieke Metals, Inc.

8-oxo-8-(2-trifluoromethylphenyl)octanoic acid

  • MDL number MFCD02260915
  • Cas Number : 898791-04-3
  • Molecular Weight : 302.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7139F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7139F
Availability details for
Quantity
  • Availability
  • Add to Cart