RIE156329915 Rieke Metals, Inc.

6-Oxo-6-(3-trifluoromethylphenyl)hexanoic acid

  • MDL number MFCD02260918
  • Cas Number : 502651-11-8
  • Molecular Weight : 274.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7140D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7140D
Availability details for
Quantity
  • Availability
  • Add to Cart