RIE156329916 Rieke Metals, Inc.

7-oxo-7-(3-trifluoromethylphenyl)heptanoic acid

  • MDL number MFCD02260919
  • Cas Number : 502651-28-7
  • Molecular Weight : 288.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7140E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7140E
Availability details for
Quantity
  • Availability
  • Add to Cart