RIE156329917 Rieke Metals, Inc.

8-oxo-8-(3-trifluoromethylphenyl)octanoic acid

  • MDL number MFCD02260920
  • Cas Number : 898791-07-6
  • Molecular Weight : 302.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7140F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7140F
Availability details for
Quantity
  • Availability
  • Add to Cart