RIE156329920 Rieke Metals, Inc.

7-Oxo-7-(4-trifluoromethylphenyl)heptanoic acid

  • MDL number MFCD02260924
  • Cas Number : 502651-46-9
  • Molecular Weight : 288.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7141E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7141E
Availability details for
Quantity
  • Availability
  • Add to Cart