RIE156329921 Rieke Metals, Inc.

5-(2,3-dichlorophenyl)-5-oxovaleric acid

  • MDL number MFCD02260927
  • Cas Number : 898791-10-1
  • Molecular Weight : 261.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7142C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7142C
Availability details for
Quantity
  • Availability
  • Add to Cart