RIE156329922 Rieke Metals, Inc.

6-(2,3-Dichlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD02260928
  • Cas Number : 898791-13-4
  • Molecular Weight : 275.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7142D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7142D
Availability details for
Quantity
  • Availability
  • Add to Cart