RIE156329926 Rieke Metals, Inc.

6-(2,4-Dichlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD02260932
  • Cas Number : 898791-22-5
  • Molecular Weight : 275.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7143D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7143D
Availability details for
Quantity
  • Availability
  • Add to Cart