RIE156329931 Rieke Metals, Inc.

6-(2,5-dichlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD02260937
  • Cas Number : 870287-01-7
  • Molecular Weight : 275.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7144D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7144D
Availability details for
Quantity
  • Availability
  • Add to Cart