RIE156329934 Rieke Metals, Inc.

6-(4-Ethylphenyl)-6-oxohexanoic acid

  • MDL number MFCD02260946
  • Cas Number : 502651-40-3
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7146D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7146D
Availability details for
Quantity
  • Availability
  • Add to Cart