RIE156329936 Rieke Metals, Inc.

8-(4-ethylphenyl)-8-oxooctanoic acid

  • MDL number MFCD02260948
  • Cas Number : 898791-35-0
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7146F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7146F
Availability details for
Quantity
  • Availability
  • Add to Cart