RIE156329937 Rieke Metals, Inc.

6-(4-Isopropylphenyl)-6-oxohexanoic acid

  • MDL number MFCD02260954
  • Cas Number : 898791-37-2
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7148D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7148D
Availability details for
Quantity
  • Availability
  • Add to Cart