RIE156329942 Rieke Metals, Inc.

8-(4-t-Butylphenyl)-8-oxooctanoic acid

  • MDL number MFCD02260965
  • Cas Number : 898791-47-4
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7150F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7150F
Availability details for
Quantity
  • Availability
  • Add to Cart