RIE156329944 Rieke Metals, Inc.

6-oxo-6-(4-n-pentylphenyl)hexanoic acid

  • MDL number MFCD02260967
  • Cas Number : 178686-75-4
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7151D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7151D
Availability details for
Quantity
  • Availability
  • Add to Cart