RIE156329946 Rieke Metals, Inc.

8-oxo-8-(4-n-pentylphenyl)octanoic acid

  • MDL number MFCD02260969
  • Cas Number : 898791-51-0
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7151F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7151F
Availability details for
Quantity
  • Availability
  • Add to Cart