RIE156329948 Rieke Metals, Inc.

6-(4-hexylphenyl)-6-oxohexanoic acid

  • MDL number MFCD02260971
  • Cas Number : 178686-77-6
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7152D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7152D
Availability details for
Quantity
  • Availability
  • Add to Cart