RIE156329949 Rieke Metals, Inc.

7-(4-hexylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD02260972
  • Cas Number : 898791-55-4
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7152E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7152E
Availability details for
Quantity
  • Availability
  • Add to Cart