RIE156329952 Rieke Metals, Inc.

6-(2-ethoxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD02260980
  • Cas Number : 898791-61-2
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7154D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7154D
Availability details for
Quantity
  • Availability
  • Add to Cart