RIE156329953 Rieke Metals, Inc.

7-(2-ethoxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD02260981
  • Cas Number : 898791-63-4
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7154E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7154E
Availability details for
Quantity
  • Availability
  • Add to Cart