RIE156329955 Rieke Metals, Inc.

5-(4-Ethoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD02260983
  • Cas Number : 34670-10-5
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7155C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7155C
Availability details for
Quantity
  • Availability
  • Add to Cart