RIE156329956 Rieke Metals, Inc.

6-(4-Ethoxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD02260984
  • Cas Number : 898791-67-8
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7155D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7155D
Availability details for
Quantity
  • Availability
  • Add to Cart