RIE156329957 Rieke Metals, Inc.

7-(4-ethoxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD02260985
  • Cas Number : 898791-69-0
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7155E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7155E
Availability details for
Quantity
  • Availability
  • Add to Cart