RIE156329963 Rieke Metals, Inc.

5-Oxo-5-(4-isopropoxyphenyl)valeric acid

  • MDL number MFCD02260992
  • Cas Number : 898791-85-0
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7157C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7157C
Availability details for
Quantity
  • Availability
  • Add to Cart