RIE156329964 Rieke Metals, Inc.

6-Oxo-6-(4-isopropoxyphenyl)hexanoic acid

  • MDL number MFCD02260993
  • Cas Number : 898791-88-3
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7157D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7157D
Availability details for
Quantity
  • Availability
  • Add to Cart