RIE156329966 Rieke Metals, Inc.

8-Oxo-8-(4-isopropoxyphenyl)octanoic acid

  • MDL number MFCD02260995
  • Cas Number : 898791-94-1
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7157F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7157F
Availability details for
Quantity
  • Availability
  • Add to Cart