RIE156329967 Rieke Metals, Inc.

5-oxo-5-(4-pentyloxyphenyl)valeric acid

  • MDL number MFCD02261000
  • Cas Number : 898791-97-4
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7159C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7159C
Availability details for
Quantity
  • Availability
  • Add to Cart