RIE156329971 Rieke Metals, Inc.

5-(4-hexyloxyphenyl)-5-oxovaleric acid

  • MDL number MFCD02261004
  • Cas Number : 898792-09-1
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7160C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7160C
Availability details for
Quantity
  • Availability
  • Add to Cart