RIE156329972 Rieke Metals, Inc.

6-(4-hexyloxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD02261005
  • Cas Number : 178686-87-8
  • Molecular Weight : 306.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7160D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7160D
Availability details for
Quantity
  • Availability
  • Add to Cart