RIE156329976 Rieke Metals, Inc.

6-(4-heptyloxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD02261009
  • Cas Number : 898792-21-7
  • Molecular Weight : 320.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7161D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7161D
Availability details for
Quantity
  • Availability
  • Add to Cart