RIE156329977 Rieke Metals, Inc.

7-(4-heptyloxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD02261010
  • Cas Number : 898792-23-9
  • Molecular Weight : 334.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7161E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7161E
Availability details for
Quantity
  • Availability
  • Add to Cart