RIE156329978 Rieke Metals, Inc.

8-(4-heptyloxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD02261011
  • Cas Number : 898792-25-1
  • Molecular Weight : 348.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7161F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7161F
Availability details for
Quantity
  • Availability
  • Add to Cart