RIE156329983 Rieke Metals, Inc.

8-(2,3-Dimethoxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD02261030
  • Cas Number : 898792-35-3
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7165F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7165F
Availability details for
Quantity
  • Availability
  • Add to Cart