RIE156329984 Rieke Metals, Inc.

6-(2,4-Dimethoxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD02261031
  • Cas Number : 898792-37-5
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7166D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7166D
Availability details for
Quantity
  • Availability
  • Add to Cart