RIE156329985 Rieke Metals, Inc.

7-(2,4-Dimethoxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD02261032
  • Cas Number : 898792-39-7
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7166E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7166E
Availability details for
Quantity
  • Availability
  • Add to Cart