RIE156329989 Rieke Metals, Inc.

4-(2,6-Dimethoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD02261037
  • Cas Number : 898792-45-5
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7168B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7168B
Availability details for
Quantity
  • Availability
  • Add to Cart