RIE156329990 Rieke Metals, Inc.

5-(2,6-Dimethoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD02261038
  • Cas Number : 898792-47-7
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7168C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7168C
Availability details for
Quantity
  • Availability
  • Add to Cart