RIE156329995 Rieke Metals, Inc.

8-(3,4-Dimethoxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD02261044
  • Cas Number : 32246-94-9
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7169F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7169F
Availability details for
Quantity
  • Availability
  • Add to Cart