RIE156329998 Rieke Metals, Inc.

7-(3,5-Dimethoxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD02261048
  • Cas Number : 52483-27-9
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7170E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7170E
Availability details for
Quantity
  • Availability
  • Add to Cart