RIE156330002 Rieke Metals, Inc.

4-[3-(N,N-Dimethylamino)phenyl]-4-oxobutyric acid

  • MDL number MFCD09801723
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7199B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7199B
Availability details for
Quantity
  • Availability
  • Add to Cart