RIE156330008 Rieke Metals, Inc.

5-[4-(N,N-Dimethylamino)phenyl]-5-oxovaleric acid

  • MDL number MFCD09801729
  • Cas Number : 139675-88-0
  • Molecular Weight : 235.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7200C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7200C
Availability details for
Quantity
  • Availability
  • Add to Cart