RIE156330010 Rieke Metals, Inc.

7-[4-(N,N-Dimethylamino)phenyl]-7-oxoheptanoic acid

  • MDL number MFCD09801731
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7200E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7200E
Availability details for
Quantity
  • Availability
  • Add to Cart