RIE156330011 Rieke Metals, Inc.

8-[4-(N,N-Dimethylamino)phenyl]-8-oxooctanoic acid

  • MDL number MFCD09801732
  • Molecular Weight : 277.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7200F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7200F
Availability details for
Quantity
  • Availability
  • Add to Cart