RIE156330013 Rieke Metals, Inc.

5-[4-(N,N-Diethylamino)phenyl]-5-oxovaleric acid

  • MDL number MFCD09801734
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7201C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7201C
Availability details for
Quantity
  • Availability
  • Add to Cart