RIE156330016 Rieke Metals, Inc.

8-[4-(N,N-Diethylamino)phenyl]-8-oxooctanoic acid

  • MDL number MFCD09801737
  • Molecular Weight : 305.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7201F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7201F
Availability details for
Quantity
  • Availability
  • Add to Cart