RIE156330019 Rieke Metals, Inc.

6-(2-Chloro-4-fluorophenyl)-6-oxohexanoic acid

  • MDL number MFCD09801740
  • Molecular Weight : 258.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7202D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7202D
Availability details for
Quantity
  • Availability
  • Add to Cart