RIE156330020 Rieke Metals, Inc.

7-(2-Chloro-4-fluorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD09801741
  • Molecular Weight : 272.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7202E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7202E
Availability details for
Quantity
  • Availability
  • Add to Cart