RIE156330021 Rieke Metals, Inc.

8-(2-Chloro-4-fluorophenyl)-8-oxooctanoic acid

  • MDL number MFCD09801742
  • Molecular Weight : 286.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7202F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7202F
Availability details for
Quantity
  • Availability
  • Add to Cart