RIE156330022 Rieke Metals, Inc.

4-(4-Chloro-2-fluorophenyl)-4-oxobutyric acid

  • MDL number MFCD09801743
  • Molecular Weight : 230.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7203B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7203B
Availability details for
Quantity
  • Availability
  • Add to Cart