RIE156330027 Rieke Metals, Inc.

4-(4-Chloro-2-methoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD09801748
  • Cas Number : 15572-02-8
  • Molecular Weight : 242.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7204B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7204B
Availability details for
Quantity
  • Availability
  • Add to Cart